Product Details

SNP ID
rs7918085
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:96841703 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAGAAGATGTTGTGAACACCAGTA[C/T]TATTTCCCATGATAATTGCTATTAT
Phenotype
MIM: 607698
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LCOR PubMed Links

Gene Details

Gene
LCOR
Gene Name
ligand dependent nuclear receptor corepressor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170765.1 Intron NP_001164236.1
NM_001170766.1 Intron NP_001164237.1
NM_032440.3 Intron NP_115816.2
XM_006718033.3 Intron XP_006718096.1
XM_011540277.2 Intron XP_011538579.1
XM_017016784.1 Intron XP_016872273.1
XM_017016785.1 Intron XP_016872274.1
XM_017016786.1 Intron XP_016872275.1
XM_017016787.1 Intron XP_016872276.1
XM_017016788.1 Intron XP_016872277.1
Gene
LOC102723665
Gene Name
uncharacterized LOC102723665
There are no transcripts associated with this gene.

View Full Product Details