Product Details

SNP ID
rs7992622
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:28138074 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGGCGCCAGATCCGCGTCCGGAAC[C/G]CCACCGAGAGCCGCCGGTCCAACCG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PAN3 PubMed Links

Gene Details

Gene
PAN3
Gene Name
PAN3 poly(A) specific ribonuclease subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175854.7 Intron NP_787050.6
XM_005266332.3 Intron XP_005266389.1
XM_005266333.3 Intron XP_005266390.1
XM_005266334.1 Intron XP_005266391.1
XM_011535032.2 Intron XP_011533334.1
XM_011535033.2 Intron XP_011533335.1
XM_011535034.2 Intron XP_011533336.1
XM_017020528.1 Intron XP_016876017.1
XM_017020529.1 Intron XP_016876018.1
XM_017020530.1 Intron XP_016876019.1
XM_017020531.1 Intron XP_016876020.1
XM_017020532.1 Intron XP_016876021.1
XM_017020533.1 Intron XP_016876022.1
Gene
PAN3-AS1
Gene Name
PAN3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details