Product Details

SNP ID
rs8001271
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:75553669 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTTGTATAGTTCTAATACCATCA[A/G]TTGTCACATAGATGACCCCATGTTC
Phenotype
MIM: 612377 MIM: 603090
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COMMD6 PubMed Links

Gene Details

Gene
COMMD6
Gene Name
COMM domain containing 6
There are no transcripts associated with this gene.

Gene
UCHL3
Gene Name
ubiquitin C-terminal hydrolase L3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270952.1 Intron NP_001257881.1
NM_006002.4 Intron NP_005993.1
XM_011535212.1 Intron XP_011533514.1
XM_011535213.2 Intron XP_011533515.1
XM_011535214.2 Intron XP_011533516.1
XM_017020725.1 Intron XP_016876214.1
XM_017020726.1 Intron XP_016876215.1
XM_017020727.1 Intron XP_016876216.1

View Full Product Details