Product Details

SNP ID
rs193302876
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:70598475 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTGGATGAAGGTCACCTCCAGCT[G/A]GGCCAGGCCCCTCTGCCGAGGGCAG
Phenotype
MIM: 170280
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
PRF1 PubMed Links

Gene Details

Gene
PRF1
Gene Name
perforin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083116.1 1407 Nonsense Mutation CAG,TAG Q416* NP_001076585.1
NM_005041.4 1407 Nonsense Mutation CAG,TAG Q416* NP_005032.2

View Full Product Details