Product Details

SNP ID
rs199759966
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:97767621 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTCCTTATTCTTCTTGTCCCAGC[A/G]GTAGACGGCCATGAGCAGCAGCTGT
Phenotype
MIM: 604158 MIM: 610243
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SFRP5 PubMed Links

Gene Details

Gene
SFRP5
Gene Name
secreted frizzled related protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003015.3 1013 Missense Mutation CGC,TGC R283C NP_003006.2
Gene
ZFYVE27
Gene Name
zinc finger FYVE-type containing 27
There are no transcripts associated with this gene.

View Full Product Details