Product Details
- SNP ID
-
rs199549238
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:59793018 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCATGGGTGGCGGAGGTGGAGGCG[C/G]AGGTGGCTGGACTGGGGTCTGTGTG
- Phenotype
-
MIM: 601985
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1053265] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CCDC6
- Gene Name
- coiled-coil domain containing 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005436.4 |
1556 |
Missense Mutation |
CCG,GCG |
P442A |
NP_005427.2 |
View Full Product Details