Product Details

SNP ID
rs199810842
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:100988736 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGATCAGGAGGAGGTTCAGCTG[A/G]CTGATACAATGTTTGGCCTTACCAA
Phenotype
MIM: 606075 MIM: 610454 MIM: 611848
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C10orf2 PubMed Links

Gene Details

Gene
C10orf2
Gene Name
chromosome 10 open reading frame 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163812.1 700 Missense Mutation ACT,GCT T176A NP_001157284.1
NM_001163813.1 700 Intron NP_001157285.1
NM_001163814.1 700 Intron NP_001157286.1
NM_021830.4 700 Missense Mutation ACT,GCT T176A NP_068602.2
XM_011539975.2 700 Intron XP_011538277.1
XM_017016437.1 700 UTR 5 XP_016871926.1
Gene
LZTS2
Gene Name
leucine zipper tumor suppressor 2
There are no transcripts associated with this gene.

Gene
MRPL43
Gene Name
mitochondrial ribosomal protein L43
There are no transcripts associated with this gene.

Gene
SEMA4G
Gene Name
semaphorin 4G
There are no transcripts associated with this gene.

View Full Product Details