Product Details

SNP ID
rs199603225
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:89306214 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAGTTAATCCAGCAAGAGCATGC[C/T]GACCAGGCAGAAATCAGAAGTCTGG
Phenotype
MIM: 147040
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IFIT2 PubMed Links

Gene Details

Gene
IFIT2
Gene Name
interferon induced protein with tetratricopeptide repeats 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001547.4 433 Silent Mutation GCC,GCT A86A NP_001538.4
Gene
LOC105378419
Gene Name
uncharacterized LOC105378419
There are no transcripts associated with this gene.

View Full Product Details