Product Details

SNP ID
rs199720543
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:123137096 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGCTGAGAGACTCCTCCCCCGCC[G/T]CCGGCACAGACCGCGACTCTCCGGA
Phenotype
MIM: 600647 MIM: 613380
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HMX2 PubMed Links

Gene Details

Gene
HMX2
Gene Name
H6 family homeobox 2
There are no transcripts associated with this gene.

Gene
HMX3
Gene Name
H6 family homeobox 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105574.1 439 Missense Mutation GCC,TCC A147S NP_001099044.1

View Full Product Details