Product Details

SNP ID
rs199722976
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:122276809 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGCAGCACAAAGCTCGGCTGGAA[C/T]GCCGGGTCACTGGCTCAACCAACCG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BTBD16 PubMed Links

Gene Details

Gene
BTBD16
Gene Name
BTB domain containing 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318189.1 298 Missense Mutation CGC,TGC R14C NP_001305118.1
NM_144587.3 298 Missense Mutation CGC,TGC R13C NP_653188.2
XM_011539239.2 298 Missense Mutation CGC,TGC R14C XP_011537541.1
XM_011539240.2 298 Missense Mutation CGC,TGC R14C XP_011537542.1
XM_011539241.2 298 Missense Mutation CGC,TGC R14C XP_011537543.1
XM_011539242.2 298 Missense Mutation CGC,TGC R14C XP_011537544.1
XM_017015636.1 298 Missense Mutation CGC,TGC R14C XP_016871125.1
XM_017015637.1 298 Intron XP_016871126.1

View Full Product Details