Product Details

SNP ID
rs201294595
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:117197709 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGAATCTTGAACTGCAGTGAAA[A/C]AGGTAGGTGCCTCACCTGGACAGGG
Phenotype
MIM: 613655
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KCNK18 PubMed Links

Gene Details

Gene
KCNK18
Gene Name
potassium two pore domain channel subfamily K member 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181840.1 221 Missense Mutation AAA,ACA K74T NP_862823.1

View Full Product Details