Product Details
- SNP ID
-
hCV189607762
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:46549430 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGGACTGCATGACAGCCCGCAGT[G/A]GCCCCCTCCGGCCCTCCACAGACAG
- Phenotype
-
MIM: 611240
- Polymorphism
- G/A, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GPRIN2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3127824] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GPRIN2
- Gene Name
- G protein regulated inducer of neurite outgrowth 2
There are no transcripts associated with this gene.
- Gene
- LOC107984026
- Gene Name
- arf-GAP with GTPase, ANK repeat and PH domain-containing protein 4-like
There are no transcripts associated with this gene.
View Full Product Details