Product Details
- SNP ID
-
rs201111934
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:114044525 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTGGGAGTACGGCTCCTTCTTCTG[C/T]GAGCTGTGGACCTCAGTGGACGTGC
- Phenotype
-
MIM: 109630
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ADRB1
PubMed Links
Gene Details
- Gene
- ADRB1
- Gene Name
- adrenoceptor beta 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000684.2 |
479 |
Silent Mutation |
TGC,TGT |
C131C |
NP_000675.1 |
View Full Product Details