Product Details
- SNP ID
-
rs201305110
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:100048810 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACAGTTACTGTCTCTGGGTCATAC[C/G]CAGGTGCTGTGGCACTAACAGTGTA
- Phenotype
-
MIM: 603103
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CPN1
PubMed Links
Gene Details
- Gene
- CPN1
- Gene Name
- carboxypeptidase N subunit 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001308.2 |
1438 |
Missense Mutation |
GCG,GGG |
A393G |
NP_001299.1 |
View Full Product Details