Product Details

SNP ID
rs201328071
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:89709382 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCTGGATTCACAGACTGTTGTGC[C/T]GAAAGAGCCTCAATGCATCCTTGGT
Phenotype
MIM: 605498
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIF20B PubMed Links

Gene Details

Gene
KIF20B
Gene Name
kinesin family member 20B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284259.1 364 Missense Mutation CCG,CTG P91L NP_001271188.1
NM_016195.3 364 Missense Mutation CCG,CTG P91L NP_057279.2

View Full Product Details