Product Details

SNP ID
rs201142410
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:133555392 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTCTTCCTCTTGTGTGCTCTGGG[A/C]TTGGGCAGGGACTTGCATTCCATGC
Phenotype
MIM: 611486
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SYCE1 PubMed Links

Gene Details

Gene
SYCE1
Gene Name
synaptonemal complex central element protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143763.1 982 Missense Mutation GCC,TCC A293S NP_001137235.1
NM_001143764.1 982 Missense Mutation GCC,TCC A293S NP_001137236.1
NM_130784.2 982 Missense Mutation GCC,TCC A257S NP_570140.1

View Full Product Details