Product Details

SNP ID
rs201361952
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:96981433 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTAGAAAGTCTAATTACAGTAAAA[A/G]TGGCAGCTGAGAATAGTGAGGAAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C10orf12 PubMed Links

Gene Details

Gene
C10orf12
Gene Name
chromosome 10 open reading frame 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015652.2 150 Missense Mutation ATG,GTG M15V NP_056467.2

View Full Product Details