Product Details

SNP ID
rs200510933
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:121963287 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAATCCCAAGATTCTTTCTACTTC[C/T]TTCTCTGTTGCTTCTTGATGAGATT
Phenotype
MIM: 612987
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NSMCE4A PubMed Links

Gene Details

Gene
NSMCE4A
Gene Name
NSE4 homolog A, SMC5-SMC6 complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167865.1 858 Silent Mutation AAA,AAG K265K NP_001161337.1
NM_017615.2 858 Silent Mutation AAA,AAG K265K NP_060085.2
XM_005269929.4 858 Silent Mutation AAA,AAG K106K XP_005269986.1
XM_005269930.3 858 Silent Mutation AAA,AAG K106K XP_005269987.1
XM_011539910.2 858 Silent Mutation AAA,AAG K106K XP_011538212.1
XM_017016359.1 858 Silent Mutation AAA,AAG K106K XP_016871848.1

View Full Product Details