Product Details

SNP ID
rs200280119
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:91908935 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCACTCAGCAGCAGCAGCAGCGAC[A/G]GCGACAGCGACGCTCGCAGCTTCGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FGFBP3 PubMed Links

Gene Details

Gene
FGFBP3
Gene Name
fibroblast growth factor binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152429.4 217 Missense Mutation CCG,CTG P12L NP_689642.3

View Full Product Details