Product Details

SNP ID
rs200420117
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:89214900 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGGGCATCCAGGCCCCAAATGAA[A/G]TCAAGATGCTCCCATTCCGGAATGC
Phenotype
MIM: 604551 MIM: 613497
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CH25H PubMed Links

Gene Details

Gene
CH25H
Gene Name
cholesterol 25-hydroxylase
There are no transcripts associated with this gene.

Gene
LIPA
Gene Name
lipase A, lysosomal acid type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000235.3 1432 Silent Mutation GAC,GAT D376D NP_000226.2
NM_001127605.2 1432 Silent Mutation GAC,GAT D376D NP_001121077.1
NM_001288979.1 1432 Silent Mutation GAC,GAT D260D NP_001275908.1

View Full Product Details