Product Details
- SNP ID
-
rs200592713
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:111078579 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATCGAGAAGAAGGGCGGCGGCGGC[A/G]GCCCGCAGCCGGCCGAGCCGCGCTG
- Phenotype
-
MIM: 104210
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ADRA2A
PubMed Links
Gene Details
- Gene
- ADRA2A
- Gene Name
- adrenoceptor alpha 2A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000681.3 |
1548 |
Missense Mutation |
AGC,GGC |
S195G |
NP_000672.3 |
View Full Product Details