Product Details

SNP ID
rs200574063
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:75105790 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCACCGCCTGGCGCAGGGACAGC[C/T]GCTGGTGCAGCATGAGGTAGGCCAG
Phenotype
MIM: 613191 MIM: 611575
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DUSP13 PubMed Links

Gene Details

Gene
DUSP13
Gene Name
dual specificity phosphatase 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007271.1 501 Missense Mutation CAG,CGG Q149R NP_001007272.1
NM_001007272.1 501 Intron NP_001007273.1
NM_001007273.1 501 Intron NP_001007274.1
NM_001320842.1 501 Intron NP_001307771.1
NM_001320843.1 501 Intron NP_001307772.1
NM_016364.3 501 Intron NP_057448.3
XM_005269883.3 501 Silent Mutation GCA,GCG A76A XP_005269940.1
XM_005269884.4 501 Intron XP_005269941.3
XM_005269887.1 501 Intron XP_005269944.1
XM_005269890.1 501 Intron XP_005269947.1
XM_011539853.1 501 UTR 5 XP_011538155.1
XM_011539854.2 501 Intron XP_011538156.1
XM_011539855.1 501 Intron XP_011538157.1
XM_011539856.2 501 Intron XP_011538158.1
XM_017016313.1 501 Intron XP_016871802.1
XM_017016314.1 501 Silent Mutation GCA,GCG A76A XP_016871803.1
Gene
SAMD8
Gene Name
sterile alpha motif domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174156.1 501 Intron NP_001167627.1
NM_144660.2 501 Intron NP_653261.1
XM_005269541.4 501 Intron XP_005269598.1
XM_011539311.1 501 Intron XP_011537613.1
XM_011539312.2 501 Intron XP_011537614.1
XM_017015738.1 501 Intron XP_016871227.1
XM_017015739.1 501 Intron XP_016871228.1

View Full Product Details