Product Details

SNP ID
rs200404406
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:125823584 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCGTCCAGGTCTAAGCGGCGTGC[A/C]GTGGAAAGTGGGGTTCCGCAGCCGC
Phenotype
MIM: 611883 MIM: 606938
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BCCIP PubMed Links

Gene Details

Gene
BCCIP
Gene Name
BRCA2 and CDKN1A interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016567.3 50 Silent Mutation GCA,GCC A9A NP_057651.1
NM_078468.2 50 Silent Mutation GCA,GCC A9A NP_510868.1
NM_078469.2 50 Silent Mutation GCA,GCC A9A NP_510869.1
Gene
MIR4484
Gene Name
microRNA 4484
There are no transcripts associated with this gene.

Gene
UROS
Gene Name
uroporphyrinogen III synthase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000375.2 50 Intron NP_000366.1
NM_001324036.1 50 Intron NP_001310965.1
NM_001324037.1 50 Intron NP_001310966.1
NM_001324038.1 50 Intron NP_001310967.1
NM_001324039.1 50 Intron NP_001310968.1
XM_005270140.4 50 Intron XP_005270197.1
XM_011540127.1 50 Intron XP_011538429.1
XM_017016610.1 50 Intron XP_016872099.1
XM_017016611.1 50 Intron XP_016872100.1
XM_017016612.1 50 Intron XP_016872101.1

View Full Product Details