Product Details

SNP ID
rs200290108
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:102484173 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGCACTTACTTCTTTTATGGCC[C/T]TGACAATCTCAAACTCAGAGGATGA
Phenotype
MIM: 605143
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACTR1A PubMed Links

Gene Details

Gene
ACTR1A
Gene Name
ARP1 actin-related protein 1 homolog A, centractin alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005736.3 752 Missense Mutation AAG,AGG K215R NP_005727.1
Gene
MFSD13A
Gene Name
major facilitator superfamily domain containing 13A
There are no transcripts associated with this gene.

View Full Product Details