Product Details

SNP ID
rs200480512
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:122932545 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACAAAGCAATCTTATCATCAATG[C/T]GCTCCTGGAGTTCATGTATTCGCTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C10orf88 PubMed Links

Gene Details

Gene
C10orf88
Gene Name
chromosome 10 open reading frame 88
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024942.3 1445 Missense Mutation CAC,CGC H407R NP_079218.2

View Full Product Details