Product Details

SNP ID
rs200362645
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:72338297 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGATAGGCATCAGCTGCACAAA[C/T]ACCCCTAGCCCGCCCTGGAGGGAAG
Phenotype
MIM: 608376
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJB12 PubMed Links

Gene Details

Gene
DNAJB12
Gene Name
DnaJ heat shock protein family (Hsp40) member B12
There are no transcripts associated with this gene.

View Full Product Details