Product Details

SNP ID
rs200773323
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:24984824 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTCTTCTTCCAGCCTCTGCTTGC[A/G]GATCTTCTTTGGTATAGAATCTATA
Phenotype
MIM: 611025 MIM: 611260
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ENKUR PubMed Links

Gene Details

Gene
ENKUR
Gene Name
enkurin, TRPC channel interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270383.1 715 Missense Mutation CGC,TGC R164C NP_001257312.1
NM_145010.3 715 Missense Mutation CGC,TGC R226C NP_659447.1
XM_006717389.1 715 Missense Mutation CGC,TGC R226C XP_006717452.1
XM_006717390.1 715 Missense Mutation CGC,TGC R130C XP_006717453.1
XM_011519348.1 715 Missense Mutation CGC,TGC R226C XP_011517650.1
XM_017015830.1 715 Missense Mutation CGC,TGC R130C XP_016871319.1
Gene
THNSL1
Gene Name
threonine synthase like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024838.4 715 Intron NP_079114.3
XM_005252597.2 715 Intron XP_005252654.1
XM_017016665.1 715 Intron XP_016872154.1

View Full Product Details