Product Details

SNP ID
rs200789852
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:70598467 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCCATGCTTGGATGAAGGTCAC[C/G]TCCAGCTGGGCCAGGCCCCTCTGCC
Phenotype
MIM: 170280
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PRF1 PubMed Links

Gene Details

Gene
PRF1
Gene Name
perforin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083116.1 1415 Silent Mutation GAC,GAG D418E NP_001076585.1
NM_005041.4 1415 Silent Mutation GAC,GAG D418E NP_005032.2

View Full Product Details