Product Details

SNP ID
rs199964528
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:70301426 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTGATGAGCGGCGGGGCGATCAC[A/G]CGCACCTCGGCGTTGAGTGGGTTGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRC20 PubMed Links

Gene Details

Gene
LRRC20
Gene Name
leucine rich repeat containing 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278211.1 643 Silent Mutation CGC,CGT R161R NP_001265140.1
NM_001278212.1 643 Silent Mutation CGC,CGT R161R NP_001265141.1
NM_001278213.1 643 Silent Mutation CGC,CGT R111R NP_001265142.1
NM_001278214.1 643 Silent Mutation CGC,CGT R92R NP_001265143.1
NM_018205.3 643 Silent Mutation CGC,CGT R105R NP_060675.1
NM_018239.3 643 Silent Mutation CGC,CGT R111R NP_060709.2
NM_207119.2 643 Silent Mutation CGC,CGT R161R NP_997002.1
XM_017016374.1 643 Silent Mutation CGC,CGT R161R XP_016871863.1
XM_017016375.1 643 Silent Mutation CGC,CGT R161R XP_016871864.1

View Full Product Details