Product Details

SNP ID
rs199940981
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:101847595 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTGTGTGGAAGGCCAGGTTTCTC[C/T]GGACGTTGGTGCTAATGGATCGAAC
Phenotype
MIM: 604661
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C10orf76 PubMed Links

Gene Details

Gene
C10orf76
Gene Name
chromosome 10 open reading frame 76
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024541.2 2150 Missense Mutation CAG,CGG Q668R NP_078817.2
XM_006717969.1 2150 Missense Mutation CAG,CGG Q598R XP_006718032.1
XM_011540151.2 2150 Missense Mutation CAG,CGG Q668R XP_011538453.1
XM_011540152.2 2150 Missense Mutation CAG,CGG Q646R XP_011538454.1
XM_011540153.2 2150 Missense Mutation CAG,CGG Q634R XP_011538455.1
XM_011540154.2 2150 Missense Mutation CAG,CGG Q612R XP_011538456.1
XM_011540156.2 2150 Missense Mutation CAG,CGG Q494R XP_011538458.1
XM_011540157.2 2150 Missense Mutation CAG,CGG Q617R XP_011538459.1
XM_017016635.1 2150 Intron XP_016872124.1
XM_017016636.1 2150 Intron XP_016872125.1
Gene
KCNIP2
Gene Name
potassium voltage-gated channel interacting protein 2
There are no transcripts associated with this gene.

View Full Product Details