Product Details
- SNP ID
-
rs200020721
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:73424560 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTCCAGTTATGATCTATTTAAATC[C/T]CGCCGTCCACTTTCTCTTCTAATTG
- Phenotype
-
MIM: 614773
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MSS51
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs61738877] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MSS51
- Gene Name
- MSS51 mitochondrial translational activator
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001024593.1 |
1442 |
Missense Mutation |
GAG,GGG |
E459G |
NP_001019764.1 |
View Full Product Details