Product Details

SNP ID
rs200072558
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:119651778 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGATCAAGATCGACCCGCAGACC[A/G]GCTGGCCCTTCTTCGTGGACCACAA
Phenotype
MIM: 603883
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
BAG3 PubMed Links

Gene Details

Gene
BAG3
Gene Name
BCL2 associated athanogene 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004281.3 409 Missense Mutation AGC,GGC S35G NP_004272.2
XM_005270287.1 409 Missense Mutation AGC,GGC S35G XP_005270344.1

View Full Product Details