Product Details

SNP ID
rs199932152
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:133326956 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATGATCAGCACGCAGCCCAGTAG[C/T]GCCATGGCGAAGGCGATCATCCGTG
Phenotype
MIM: 604647
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CALY PubMed Links

Gene Details

Gene
CALY
Gene Name
calcyon neuron specific vesicular protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321617.1 1584 UTR 5 NP_001308546.1
NM_015722.3 1584 Silent Mutation NP_056537.1
XM_005252685.4 1584 Silent Mutation XP_005252742.2
XM_011539836.2 1584 Silent Mutation XP_011538138.1
XM_011539838.2 1584 Silent Mutation XP_011538140.1

View Full Product Details