Product Details

SNP ID
rs199955463
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:122912768 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGTGTCTTTTCTTAAGAGCTGC[A/G]AAGGACCCTGATGCTGTGGCTGTAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM24A PubMed Links

Gene Details

Gene
FAM24A
Gene Name
family with sequence similarity 24 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029888.1 261 Silent Mutation GCA,GCG A44A NP_001025059.1
XM_017015638.1 261 Silent Mutation GCA,GCG A44A XP_016871127.1
XM_017015639.1 261 Silent Mutation GCA,GCG A44A XP_016871128.1

View Full Product Details