Product Details

SNP ID
rs199861252
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:58388206 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCCATAGATGCAAAAACTACAGA[A/G]CTAATTAGAAGAATTGCCCAGCGTT
Phenotype
MIM: 600438
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TFAM PubMed Links

Gene Details

Gene
TFAM
Gene Name
transcription factor A, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270782.1 642 Silent Mutation GAA,GAG E79E NP_001257711.1
NM_003201.2 642 Silent Mutation GAA,GAG E79E NP_003192.1
XM_011540120.2 642 Silent Mutation GAA,GAG E79E XP_011538422.1
XM_011540121.2 642 Silent Mutation GAA,GAG E79E XP_011538423.1

View Full Product Details