Product Details

SNP ID
rs199957571
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:72275075 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGATCCAGGGGCTGTTTAGCTCC[A/G]CCAACTCTCCCTTCCTCCCTGGCTT
Phenotype
MIM: 607729
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DDIT4 PubMed Links

Gene Details

Gene
DDIT4
Gene Name
DNA damage inducible transcript 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019058.2 788 Missense Mutation ACC,GCC T196A NP_061931.1

View Full Product Details