Product Details

SNP ID
rs199957632
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:133352015 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAACTGTGCCAGCAGGGCCTGGG[A/G]CGCCCGTGTGGTGGAGCCTCCGGAG
Phenotype
MIM: 609776
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FUOM PubMed Links

Gene Details

Gene
FUOM
Gene Name
fucose mutarotase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098483.2 215 Intron NP_001091953.1
NM_001301827.1 215 Intron NP_001288756.1
NM_001301828.1 215 Intron NP_001288757.1
NM_198472.2 215 Intron NP_940874.2
XM_011539670.2 215 Intron XP_011537972.1
XM_011539671.2 215 Intron XP_011537973.1
XM_011539672.2 215 Intron XP_011537974.1
XM_011539673.2 215 Intron XP_011537975.1
XM_011539674.2 215 Intron XP_011537976.1
XM_011539675.2 215 Intron XP_011537977.1
XM_017016094.1 215 Intron XP_016871583.1
Gene
PRAP1
Gene Name
proline rich acidic protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145201.1 215 Missense Mutation GAC,GGC D46G NP_001138673.1
NM_145202.4 215 Missense Mutation GAC,GGC D46G NP_660203.3

View Full Product Details