Product Details

SNP ID
rs201791108
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:97590132 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCACGGTTCACATAGAGACCTTC[A/C]CCACGAGGCACGGAGAAGTGGGCTC
Phenotype
MIM: 610734 MIM: 613597
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD2 PubMed Links

Gene Details

Gene
ANKRD2
Gene Name
ankyrin repeat domain 2
There are no transcripts associated with this gene.

Gene
C10orf62
Gene Name
chromosome 10 open reading frame 62
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009997.2 440 Missense Mutation ACC,CCC T79P NP_001009997.2
Gene
HOGA1
Gene Name
4-hydroxy-2-oxoglutarate aldolase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134670.1 440 Intron NP_001128142.1
NM_138413.3 440 Intron NP_612422.2

View Full Product Details