Product Details

SNP ID
rs201848733
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:132447930 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGCGGCCTCCGCACTCCAGTGGC[C/T]CAGACCTGGCTGAGGCAGCTCCCGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C10orf91 PubMed Links

Gene Details

Gene
C10orf91
Gene Name
chromosome 10 open reading frame 91
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173541.2 347 Missense Mutation CCA,TCA P103S NP_775812.1
Gene
LOC105378568
Gene Name
uncharacterized LOC105378568
There are no transcripts associated with this gene.

View Full Product Details