Product Details
- SNP ID
-
rs202040509
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:70772523 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTACTTGGAATCTTACCTGGTTTTT[C/T]GCTTTTTGTAAAAGGTGATATCTTC
- Phenotype
-
MIM: 612640
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TBATA
PubMed Links
Gene Details
- Gene
- TBATA
- Gene Name
- thymus, brain and testes associated
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001318241.1 |
1496 |
Missense Mutation |
AAA,GAA |
K322E |
NP_001305170.1 |
NM_001318242.1 |
1496 |
Missense Mutation |
AAA,GAA |
K321E |
NP_001305171.1 |
NM_001318243.1 |
1496 |
Missense Mutation |
AAA,GAA |
K320E |
NP_001305172.1 |
NM_152710.3 |
1496 |
Missense Mutation |
AAA,GAA |
K321E |
NP_689923.3 |
XM_017015845.1 |
1496 |
Silent Mutation |
GCA,GCG |
A384A |
XP_016871334.1 |
XM_017015846.1 |
1496 |
Silent Mutation |
GCA,GCG |
A384A |
XP_016871335.1 |
XM_017015847.1 |
1496 |
Silent Mutation |
GCA,GCG |
A384A |
XP_016871336.1 |
XM_017015848.1 |
1496 |
Missense Mutation |
CAA,CGA |
Q369R |
XP_016871337.1 |
XM_017015849.1 |
1496 |
Silent Mutation |
GCA,GCG |
A384A |
XP_016871338.1 |
XM_017015850.1 |
1496 |
Intron |
|
|
XP_016871339.1 |
XM_017015851.1 |
1496 |
Intron |
|
|
XP_016871340.1 |
XM_017015852.1 |
1496 |
Intron |
|
|
XP_016871341.1 |
XM_017015853.1 |
1496 |
Missense Mutation |
AAA,GAA |
K353E |
XP_016871342.1 |
XM_017015854.1 |
1496 |
Missense Mutation |
CAA,CGA |
Q369R |
XP_016871343.1 |
XM_017015855.1 |
1496 |
Missense Mutation |
CAA,CGA |
Q369R |
XP_016871344.1 |
XM_017015856.1 |
1496 |
Missense Mutation |
AAA,GAA |
K353E |
XP_016871345.1 |
XM_017015857.1 |
1496 |
Missense Mutation |
AAA,GAA |
K353E |
XP_016871346.1 |
XM_017015858.1 |
1496 |
Intron |
|
|
XP_016871347.1 |
XM_017015859.1 |
1496 |
Intron |
|
|
XP_016871348.1 |
XM_017015860.1 |
1496 |
Missense Mutation |
CAA,CGA |
Q277R |
XP_016871349.1 |
XM_017015861.1 |
1496 |
Intron |
|
|
XP_016871350.1 |
XM_017015862.1 |
1496 |
Intron |
|
|
XP_016871351.1 |
View Full Product Details