Product Details

SNP ID
rs202073747
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:84141607 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAAGAATTCCATCACGAAGAATC[A/G]ATGGCTGTTAACACCTAGCAGGGTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GHITM PubMed Links

Gene Details

Gene
GHITM
Gene Name
growth hormone inducible transmembrane protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014394.2 311 Missense Mutation CAA,CGA Q36R NP_055209.2

View Full Product Details