Product Details

SNP ID
rs201811839
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:26216824 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCAAAGCCGATGGCATCTCCGGG[C/T]TCTGGCTTTTGGTCTTTCGGGTCGG
Phenotype
MIM: 138275 MIM: 606808
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GAD2 PubMed Links

Gene Details

Gene
GAD2
Gene Name
glutamate decarboxylase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000818.2 518 Silent Mutation GGC,GGT G5G NP_000809.1
NM_001134366.1 518 Silent Mutation GGC,GGT G5G NP_001127838.1
Gene
MYO3A
Gene Name
myosin IIIA
There are no transcripts associated with this gene.

View Full Product Details