Product Details

SNP ID
rs201628418
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:102066369 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTATTGCAGTCCCACGGTGGTACG[C/T]GGGCTGTGGGCACCCTGCAGGAGGC
Phenotype
MIM: 607522
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HPS6 PubMed Links

Gene Details

Gene
HPS6
Gene Name
HPS6, biogenesis of lysosomal organelles complex 2 subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024747.5 1003 Missense Mutation CGG,TGG R299W NP_079023.2

View Full Product Details