Product Details
- SNP ID
-
rs201538793
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:124992705 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTTCCTTTTCTCACCTGTATGTGG[C/T]ACCATTGAGCTCAGGATGAATTGCT
- Phenotype
-
MIM: 602619
MIM: 611749
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CTBP2
PubMed Links
Gene Details
- Gene
- CTBP2
- Gene Name
- C-terminal binding protein 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001083914.2 |
3355 |
Intron |
|
|
NP_001077383.1 |
NM_001290214.2 |
3355 |
Intron |
|
|
NP_001277143.1 |
NM_001290215.2 |
3355 |
Missense Mutation |
ACC,GCC |
T383A |
NP_001277144.1 |
NM_001321012.1 |
3355 |
Missense Mutation |
ACC,GCC |
T383A |
NP_001307941.1 |
NM_001321013.1 |
3355 |
Missense Mutation |
ACC,GCC |
T383A |
NP_001307942.1 |
NM_001321014.1 |
3355 |
Missense Mutation |
ACC,GCC |
T383A |
NP_001307943.1 |
NM_001329.3 |
3355 |
Intron |
|
|
NP_001320.1 |
NM_022802.2 |
3355 |
Missense Mutation |
ACC,GCC |
T923A |
NP_073713.2 |
XM_005269561.2 |
3355 |
Missense Mutation |
ACC,GCC |
T383A |
XP_005269618.1 |
XM_005269564.2 |
3355 |
Missense Mutation |
ACC,GCC |
T383A |
XP_005269621.1 |
XM_005269567.2 |
3355 |
Missense Mutation |
ACC,GCC |
T383A |
XP_005269624.1 |
XM_005269568.4 |
3355 |
Missense Mutation |
ACC,GCC |
T383A |
XP_005269625.1 |
XM_005269569.2 |
3355 |
Missense Mutation |
ACC,GCC |
T383A |
XP_005269626.1 |
XM_005269571.2 |
3355 |
Missense Mutation |
ACC,GCC |
T383A |
XP_005269628.1 |
XM_005269572.3 |
3355 |
Missense Mutation |
ACC,GCC |
T383A |
XP_005269629.1 |
XM_006717642.2 |
3355 |
Missense Mutation |
ACC,GCC |
T383A |
XP_006717705.1 |
XM_011539349.2 |
3355 |
Missense Mutation |
ACC,GCC |
T451A |
XP_011537651.1 |
XM_011539351.1 |
3355 |
Missense Mutation |
ACC,GCC |
T383A |
XP_011537653.1 |
XM_011539353.1 |
3355 |
Missense Mutation |
ACC,GCC |
T383A |
XP_011537655.1 |
XM_011539354.1 |
3355 |
Missense Mutation |
ACC,GCC |
T383A |
XP_011537656.1 |
XM_011539355.1 |
3355 |
Missense Mutation |
ACC,GCC |
T383A |
XP_011537657.1 |
XM_011539357.1 |
3355 |
Missense Mutation |
ACC,GCC |
T358A |
XP_011537659.1 |
XM_011539358.1 |
3355 |
Missense Mutation |
ACC,GCC |
T358A |
XP_011537660.1 |
XM_017015756.1 |
3355 |
Missense Mutation |
ACC,GCC |
T383A |
XP_016871245.1 |
XM_017015757.1 |
3355 |
Missense Mutation |
ACC,GCC |
T383A |
XP_016871246.1 |
- Gene
- ZRANB1
- Gene Name
- zinc finger RANBP2-type containing 1
There are no transcripts associated with this gene.
View Full Product Details