Product Details

SNP ID
rs201493683
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:23104201 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAGTGAGTGGCAAAAGAAACTAA[C/T]CCCGGAGCAGTTCTACGTCACAAGA
Phenotype
MIM: 613782
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MSRB2 PubMed Links

Gene Details

Gene
MSRB2
Gene Name
methionine sulfoxide reductase B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012228.3 287 Missense Mutation ACC,ATC T59I NP_036360.3
XM_011519426.2 287 Missense Mutation ACC,ATC T59I XP_011517728.1

View Full Product Details