Product Details

SNP ID
rs201660711
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:93062146 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTGGGCGAGCACCGCCTGGTGC[C/G]CAGCCAGTGGCCGCAGAGTGCGCAC
Phenotype
MIM: 608428 MIM: 609672
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CYP26C1 PubMed Links

Gene Details

Gene
CYP26C1
Gene Name
cytochrome P450 family 26 subfamily C member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_183374.2 341 Missense Mutation CCC,CGC P114R NP_899230.2
Gene
EXOC6
Gene Name
exocyst complex component 6
There are no transcripts associated with this gene.

View Full Product Details