Product Details

SNP ID
rs201663287
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:59652856 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTGCATGTATCCCAAGAGGGCAA[A/G]GCTGCCAGCAGCAGAATAAAACCTC
Phenotype
MIM: 614242
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
SLC16A9 PubMed Links

Gene Details

Gene
SLC16A9
Gene Name
solute carrier family 16 member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323977.1 1848 Silent Mutation GCC,GCT A395A NP_001310906.1
NM_001323978.1 1848 Silent Mutation GCC,GCT A395A NP_001310907.1
NM_001323979.1 1848 Silent Mutation GCC,GCT A395A NP_001310908.1
NM_001323980.1 1848 Silent Mutation GCC,GCT A395A NP_001310909.1
NM_001323981.1 1848 Silent Mutation GCC,GCT A482A NP_001310910.1
NM_194298.2 1848 Silent Mutation GCC,GCT A482A NP_919274.1
XM_017015883.1 1848 Silent Mutation GCC,GCT A524A XP_016871372.1
XM_017015884.1 1848 Silent Mutation GCC,GCT A482A XP_016871373.1

View Full Product Details