Product Details
- SNP ID
-
hCV189712097
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:747566 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTCACCACCGTGGTGGCCGACACG[G/T]GCGACTTCCACGGTGAGGACGGCGC
- Phenotype
-
MIM: 602063
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TALDO1
PubMed Links
Gene Details
- Gene
- TALDO1
- Gene Name
- transaldolase 1
There are no transcripts associated with this gene.
View Full Product Details