Product Details
- SNP ID
-
rs199499331
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:5132067 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATATCTTGTTGACTCGGATATCTTC[A/G]GTAGCCAGCTTCACGATGGCCATGT
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR52A5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2472530] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR52A5
- Gene Name
- olfactory receptor family 52 subfamily A member 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001005160.2 |
576 |
Silent Mutation |
ACC,ACT |
T192T |
NP_001005160.1 |
View Full Product Details