Product Details

SNP ID
rs199568088
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:63681696 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATCTCCTCGTCGTCCTTCGGAGC[C/G]GAGCCGTCCGCGCCCGGCGGCGGCG
Phenotype
MIM: 609369 MIM: 604249
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ATL3 PubMed Links

Gene Details

Gene
ATL3
Gene Name
atlastin GTPase 3
There are no transcripts associated with this gene.

Gene
RTN3
Gene Name
reticulon 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001265589.1 337 Intron NP_001252518.1
NM_001265590.1 337 Intron NP_001252519.1
NM_001265591.1 337 Intron NP_001252520.1
NM_006054.3 337 Intron NP_006045.1
NM_201428.2 337 Intron NP_958831.1
NM_201429.2 337 Intron NP_958832.1
NM_201430.2 337 Intron NP_958833.1
XM_011544730.2 337 Intron XP_011543032.1
XM_011544731.2 337 Intron XP_011543033.1
XM_017017091.1 337 Silent Mutation GCC,GCG A20A XP_016872580.1
XM_017017092.1 337 Silent Mutation GCC,GCG A20A XP_016872581.1

View Full Product Details